siRNA Details
Gene Symbol: SYNE1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120501.1 | 2 | 202 | |||
GenBank mRNA | CK821189.1 | - | 2 | 202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000412161.1 | - | 2 | 186 | Chr. 6 - 152380546 - 152381564 [+] on Build GRCh38 |
