siRNA Details
| Gene Symbol: MAPKAPK5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC007973.2 | - | 2, 3 | 374 | ||
| BC015633.2 | - | 2, 3 | 743 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAPKAPK5-AS1| Gene Symbol: MAPKAPK5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC007973.2 | - | 2, 3 | 374 | ||
| BC015633.2 | - | 2, 3 | 743 | |||