siRNA Details
Gene Symbol: LINC00380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104057.1 | 3 | 373 | |||
GenBank mRNA | BX097335.1 | - | 3 | 373 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000442478.1 | - | 2 | 328 | Chr. 13 - 91087254 - 91089593 [-] on Build GRCh38 |
