siRNA Details
| Gene Symbol: LOC101929420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110870.1 | 5 | 1772 | |||
| GenBank mRNA | AK055794.1 | - | 5 | 1772 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000446811.1 | - | 5 | 1772 | Chr. 6 - 168242938 - 168262581 [+] on Build GRCh38 | |

LOC101929420
Genomic Map