siRNA Details
| Gene Symbol: CHL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110739.1 | 3 | 628 | |||
| GenBank mRNA | BC029457.1 | - | 3 | 629 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417612.1 | - | 3 | 628 | Chr. 3 - 363370 - 385795 [-] on Build GRCh38 | |

CHL1-AS1
Genomic Map