siRNA Details
| Gene Symbol: LOC101927043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110207.1 | 2 | 581 | |||
| GenBank mRNA | AK094343.1 | - | 2 | 473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927043| Gene Symbol: LOC101927043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110207.1 | 2 | 581 | |||
| GenBank mRNA | AK094343.1 | - | 2 | 473 | ||