siRNA Details
| Gene Symbol: LOC101927043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110207.1 | 3 | 777 | |||
| GenBank mRNA | AK094343.1 | - | 3 | 669 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419035.1 | - | 3 | 669 | Chr. 2 - 47213949 - 47344966 [-] on Build GRCh38 | |

LOC101927043
Genomic Map