siRNA Details
| Gene Symbol: LOC100506175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111906.1 | 3 | 315 | |||
| GenBank mRNA | AA044156.1 | - | 3 | 315 | ||
| CF130631.1 | - | 3 | 175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506175| Gene Symbol: LOC100506175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111906.1 | 3 | 315 | |||
| GenBank mRNA | AA044156.1 | - | 3 | 315 | ||
| CF130631.1 | - | 3 | 175 | |||