siRNA Details
| Gene Symbol: GPR50-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135300.1 | 3 | 1570 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000454196.1 | - | 3 | 1570 | Chr. X - 151175192 - 151177836 [-] on Build GRCh38 | |

GPR50-AS1
Genomic Map