siRNA Details
Gene Symbol: GPR50-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135300.1 | 3 | 1499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000454196.1 | - | 3 | 1499 | Chr. X - 151175192 - 151177836 [-] on Build GRCh38 |
