siRNA Details
Gene Symbol: LOC100996654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110040.1 | 2 | 298 | |||
GenBank mRNA | HY038517.1 | - | 2 | 299 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110040.1 | 2 | 298 | |||
GenBank mRNA | HY038517.1 | - | 2 | 299 |