siRNA Details
| Gene Symbol: LOC101927870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110141.1 | 2 | 1340 | |||
| GenBank mRNA | BC041378.2 | - | 2 | 1340 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000422488.1 | - | 2 | 1340 | Chr. 7 - 103445207 - 103514007 [+] on Build GRCh38 | |

LOC101927870
Genomic Map