siRNA Details
| Gene Symbol: LINC00412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120429.1 | 1, 2 | 559 | |||
| GenBank mRNA | BX108969.1 | - | 1, 2 | 559 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444744.1 | - | 1, 2 | 113 | Chr. 13 - 27236282 - 27236811 [-] on Build GRCh38 | |

LINC00412
Genomic Map