siRNA Details
| Gene Symbol: LOC100507506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103764.1 | 4 | 1030 | |||
| GenBank mRNA | AK092451.1 | - | 4 | 1030 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427049.2 | - | 4 | 1030 | Chr. 6 - 4136072 - 4157385 [+] on Build GRCh38 | |

LOC100507506
Genomic Map