siRNA Details
| Gene Symbol: LINC01239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038977.1 | 4 | 438 | |||
| GenBank mRNA | AK092601.1 | - | 4 | 438 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436786.1 | - | 4 | 438 | Chr. 9 - 22646200 - 22824213 [+] on Build GRCh38 | |

LINC01239
Genomic Map