siRNA Details
Gene Symbol: LINC01239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038977.1 | 6 | 2247 | |||
GenBank mRNA | AK092601.1 | - | 6 | 2253 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000436786.1 | - | 6 | 2247 | Chr. 9 - 22646200 - 22824213 [+] on Build GRCh38 |
