siRNA Details
Gene Symbol: LOC101929374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110538.1 | 2 | 567 | |||
GenBank mRNA | BI769095.1 | - | 2 | 567 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000428415.1 | - | 2 | 234 | Chr. 22 - 23434171 - 23434731 [+] on Build GRCh38 |
