siRNA Details
| Gene Symbol: LOC101929445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134479.1 | 2 | 1315 | |||
| GenBank mRNA | AK094093.1 | - | 2 | 1318 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421320.1 | - | 2 | 1315 | Chr. 10 - 42871521 - 42874060 [+] on Build GRCh38 | |

LOC101929445
Genomic Map