siRNA Details
| Gene Symbol: LINC01524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110038.1 | 1 | 14 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000428009.1 | - | 1 | 14 | Chr. 20 - 52620838 - 52650431 [+] on Build GRCh38 | |

LINC01524
Genomic Map