siRNA Details
Gene Symbol: LINC00700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040253.2 | 3 | 1720 | |||
GenBank mRNA | AK097474.1 | - | 3 | 1721 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000413603.5 | - | 3 | 1720 | Chr. 10 - 2005473 - 2014348 [-] on Build GRCh38 |
