siRNA Details
| Gene Symbol: SEMA3F-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135301.1 | 4 | 976 | |||
| GenBank mRNA | AK125500.1 | - | 4 | 976 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000437204.1 | - | 4 | 976 | Chr. 3 - 50116022 - 50156085 [-] on Build GRCh38 | |

SEMA3F-AS1
Genomic Map