siRNA Details
Gene Symbol: SEMA3F-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135301.1 | 4 | 2508 | |||
GenBank mRNA | AK125500.1 | - | 4 | 2511 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000437204.1 | - | 4 | 2508 | Chr. 3 - 50116022 - 50156085 [-] on Build GRCh38 |
