siRNA Details
| Gene Symbol: LINC00330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038433.2 | 3 | 1008 | |||
| GenBank mRNA | AK056732.1 | - | 3 | 1009 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000414562.1 | - | 3 | 1008 | Chr. 13 - 44799503 - 44809630 [-] on Build GRCh38 | |

LINC00330
Genomic Map