siRNA Details
| Gene Symbol: LOC100130548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135126.1 | 3 | 879 | |||
| GenBank mRNA | AK057520.1 | - | 3 | 879 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412181.1 | - | 3 | 879 | Chr. 9 - 134054290 - 134058805 [+] on Build GRCh38 | |

LOC100130548
Genomic Map