siRNA Details
| Gene Symbol: LOC101927378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110079.1 | 3 | 3935 | |||
| GenBank mRNA | BX647900.1 | - | 3 | 3936 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000450977.2 | - | 3 | 3935 | Chr. 7 - 84532476 - 84574843 [+] on Build GRCh38 | |

LOC101927378
Genomic Map