siRNA Details
| Gene Symbol: CNTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125798.1 | - | 2 | 2693 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451707.1 | - | 2 | 2694 | Chr. 3 - 1008135 - 1012934 [+] on Build GRCh38 | |

CNTN6
Genomic Map