siRNA Details
Gene Symbol: LOC105378614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135054.1 | 1, 2 | 125 | |||
GenBank mRNA | AI791463.1 | - | 1, 2 | 155 | ||
BX106629.1 | - | 1, 2 | 144 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000416470.1 | - | 1, 2 | 125 | Chr. 1 - 19591802 - 19596832 [-] on Build GRCh38 |
