siRNA Details
| Gene Symbol: LOC105378614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135054.1 | 1 | 74 | |||
| GenBank mRNA | AI791463.1 | - | 1 | 104 | ||
| BX106629.1 | - | 1 | 93 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000416470.1 | - | 1 | 74 | Chr. 1 - 19591802 - 19596832 [-] on Build GRCh38 | |

LOC105378614
Genomic Map