siRNA Details
Gene Symbol: THRA1/BTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131239.1 | 3 | 422 | |||
NR_131240.1 | 2 | 362 | ||||
GenBank mRNA | BG185647.1 | - | 1 | 197 | ||
BG197541.1 | - | 1 | 205 | |||
DA697253.1 | - | 3 | 394 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000421610.2 | - | 2 | 362 | Chr. 17 - 48294347 - 48307829 [+] on Build GRCh38 |
