siRNA Details
Gene Symbol: LINC01281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038968.1 | 1 | 48 | |||
GenBank mRNA | AK093678.1 | - | 1 | 49 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000448597.1 | - | 1 | 48 | Chr. X - 39304956 - 39327362 [-] on Build GRCh38 |
