siRNA Details
| Gene Symbol: LINC01281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038968.1 | 3 | 442 | |||
| GenBank mRNA | AK093678.1 | - | 3 | 443 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448597.1 | - | 3 | 442 | Chr. X - 39304956 - 39327362 [-] on Build GRCh38 | |

LINC01281
Genomic Map