siRNA Details
| Gene Symbol: LOC101929350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134584.1 | 2 | 591 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000422917.1 | - | 2 | 798 | Chr. 22 - 16746869 - 16748645 [-] on Build GRCh38 | |

LOC101929350
Genomic Map