siRNA Details
| Gene Symbol: LOC101928453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120638.1 | 1 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928453| Gene Symbol: LOC101928453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120638.1 | 1 | 410 | |||