siRNA Details
Gene Symbol: NEURL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120675.1 | 2 | 105 | |||
NR_120676.1 | 2 | 105 | ||||
GenBank mRNA | AA399019.1 | - | 1 | 56 | ||
BX089018.1 | - | 2 | 105 | |||
BX095052.1 | - | 2 | 79 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000447820.1 | - | 1 | 48 | Chr. 10 - 103511779 - 103515402 [-] on Build GRCh38 |
