siRNA Details
| Gene Symbol: LINC01372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108104.1 | 3 | 445 | |||
| GenBank mRNA | BC017910.1 | - | 3 | 446 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000430244.1 | - | 3 | 445 | Chr. 7 - 67335976 - 67340024 [+] on Build GRCh38 | |

LINC01372
Genomic Map