siRNA Details
Gene Symbol: LINC01372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108104.1 | 3 | 351 | |||
GenBank mRNA | BC017910.1 | - | 3 | 352 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000430244.1 | - | 3 | 351 | Chr. 7 - 67335976 - 67340024 [+] on Build GRCh38 |
