siRNA Details
Gene Symbol: LOC105373496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135564.1 | 3 | 428 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000425578.1 | - | 3 | 428 | Chr. 2 - 96527940 - 96532306 [-] on Build GRCh38 |
