siRNA Details
| Gene Symbol: HIPK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110725.1 | 5 | 1946 | |||
| NR_110726.1 | 3 | 470 | ||||
| GenBank mRNA | BC036361.1 | - | 5 | 1947 | ||
| BC048113.1 | - | 2 | 1150 | |||
| HY108301.1 | - | 3 | 470 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIPK1-AS1| Gene Symbol: HIPK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110725.1 | 5 | 1946 | |||
| NR_110726.1 | 3 | 470 | ||||
| GenBank mRNA | BC036361.1 | - | 5 | 1947 | ||
| BC048113.1 | - | 2 | 1150 | |||
| HY108301.1 | - | 3 | 470 | |||