siRNA Details
| Gene Symbol: HIPK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110725.1 | 1 | 254 | |||
| GenBank mRNA | BC036361.1 | - | 1 | 254 | ||
| BC048113.1 | - | 1 | 228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIPK1-AS1| Gene Symbol: HIPK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110725.1 | 1 | 254 | |||
| GenBank mRNA | BC036361.1 | - | 1 | 254 | ||
| BC048113.1 | - | 1 | 228 | |||