siRNA Details
| Gene Symbol: MSC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033651.1 | 3 | 755 | |||
| NR_033652.1 | 5 | 1350 | ||||
| GenBank mRNA | AK123315.1 | - | 5 | 710 | ||
| AK304427.1 | - | 5 | 1350 | |||
| BC094782.1 | - | 4 | 921 | |||
| BQ332273.1 | - | 1 | 294 | |||
| BX647470.1 | - | 3 | 768 | |||

MSC-AS1
Genomic Map