siRNA Details
| Gene Symbol: MSC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033652.1 | 1 | 93 | |||
| GenBank mRNA | AK304427.1 | - | 1 | 93 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000522519.5 | - | 1 | 93 | Chr. 8 - 71843123 - 72053047 [+] on Build GRCh38 | |

MSC-AS1
Genomic Map