siRNA Details
| Gene Symbol: LOC105379783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094048.1 | - | 1 | 1476 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439621.1 | - | 1 | 1476 | Chr. 1 - 53114576 - 53118609 [+] on Build GRCh38 | |

LOC105379783
Genomic Map