siRNA Details
Gene Symbol: LOC105379783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094048.1 | - | 1 | 1450 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000439621.1 | - | 1 | 1450 | Chr. 1 - 53114576 - 53118609 [+] on Build GRCh38 |
