siRNA Details
| Gene Symbol: LINC00484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DB291835.1 | - | 1 | 23 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000423719.3 | - | 1 | 51 | Chr. 9 - 91159573 - 91165576 [+] on Build GRCh38 | |

LINC00484
Genomic Map