siRNA Details
| Gene Symbol: LOC101929181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104624.1 | 3 | 842 | |||
| GenBank mRNA | BC039491.1 | - | 3 | 842 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453135.1 | - | 3 | 842 | Chr. 1 - 11609531 - 11613355 [+] on Build GRCh38 | |

LOC101929181
Genomic Map