siRNA Details
| Gene Symbol: LOC100507557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038244.1 | 4 | 801 | |||
| NR_038245.1 | 3 | 695 | ||||
| NR_038246.1 | 3 | 227 | ||||
| GenBank mRNA | AK124757.1 | - | 3 | 224 | ||
| AK311235.1 | - | 3 | 659 | |||
| BC043592.1 | - | 4 | 435 | |||
| DB465589.1 | - | 3 | 191 | |||

LOC100507557
Genomic Map