siRNA Details
| Gene Symbol: LINC00298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015405.1 | 3 | 1347 | |||
| GenBank mRNA | BC043361.1 | - | 4 | 1418 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000426969.5 | - | 4 | 1418 | Chr. 2 - 7922425 - 7979310 [-] on Build GRCh38 | |

LINC00298
Genomic Map