siRNA Details
| Gene Symbol: LOC101928663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110862.1 | 4 | 689 | |||
| NR_110863.1 | 3 | 571 | ||||
| GenBank mRNA | BC029534.1 | - | 4 | 689 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928663| Gene Symbol: LOC101928663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110862.1 | 4 | 689 | |||
| NR_110863.1 | 3 | 571 | ||||
| GenBank mRNA | BC029534.1 | - | 4 | 689 | ||