siRNA Details
| Gene Symbol: LINC00607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037195.1 | 10 | 2492 | |||
| GenBank mRNA | AK124897.1 | - | 10 | 2492 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000445174.5 | - | 10 | 2492 | Chr. 2 - 215611563 - 215843536 [-] on Build GRCh38 | |

LINC00607
Genomic Map