siRNA Details
| Gene Symbol: BHLHE40-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037903.3 | 3 | 524 | |||
| NR_125914.1 | 3 | 483 | ||||
| GenBank mRNA | AK311159.1 | - | 3 | 483 | ||
| AK311646.1 | - | 3 | 500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BHLHE40-AS1| Gene Symbol: BHLHE40-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037903.3 | 3 | 524 | |||
| NR_125914.1 | 3 | 483 | ||||
| GenBank mRNA | AK311159.1 | - | 3 | 483 | ||
| AK311646.1 | - | 3 | 500 | |||